Assay Details
| Gene Symbol: HCAR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100341.4 | NP_171956.2 | 18 - 19 | 1419 | 123 | |
| GenBank mRNA | AK117982.1 | - | 18 - 19 | 1360 | 123 | |
| BT002029.1 | - | 18 - 19 | 1384 | 123 | ||
| BT006603.1 | - | 18 - 19 | 1292 | 123 | ||
More Information
Set Membership: |
|

HCAR
Genomic Map