Assay Details
| Gene Symbol: AT1G04680 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100348.4 | NP_563715.1 | 3 - 4 | 1011 | 74 | |
| GenBank mRNA | AY065034.1 | - | 3 - 4 | 901 | 74 | |
| AY087724.1 | - | 3 - 4 | 920 | 74 | ||
More Information
Set Membership: |
|

AT1G04680
Genomic Map