Assay Details
| Gene Symbol: RCI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100405.4 | NP_172018.1 | 2 - 3 | 464 | 74 | |
| GenBank mRNA | AK227859.1 | - | 2 - 3 | 459 | 74 | |
| BT004817.1 | - | 2 - 3 | 404 | 74 | ||
| U97684.1 | - | 2 - 3 | 438 | 74 | ||
More Information
Set Membership: |
|

RCI3
Genomic Map