Assay Details
| Gene Symbol: AT1G05290 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100408.2 | NP_172021.2 | 1 - 2 | 487 | 113 | |
| GenBank mRNA | DQ446230.1 | - | 1 - 2 | 471 | 113 | |
More Information
Set Membership: |
|

AT1G05290
Genomic Map