Assay Details
| Gene Symbol: ATX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331588.1 | NP_001321564.1 | 24 - 25 | 3572 | 116 | |
| NM_100464.7 | NP_172074.6 | 24 - 25 | 3542 | 116 | ||
More Information
Set Membership: |
|

ATX2
Genomic Map