Assay Details
| Gene Symbol: ATX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083995.4 | NP_001077464.4 | 2 - 3 | 881 | 97 | |
| NM_001331587.1 | NP_001321563.1 | 2 - 3 | 995 | 97 | ||
| NM_001331588.1 | NP_001321564.1 | 2 - 3 | 995 | 97 | ||
| NM_100464.7 | NP_172074.6 | 2 - 3 | 965 | 97 | ||
More Information
Set Membership: |
|

ATX2
Genomic Map