Assay Details
| Gene Symbol: POM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331589.1 | NP_001321426.1 | 3 - 4 | 1154 | 52 | |
| NM_100466.5 | NP_172076.1 | 3 - 4 | 869 | 52 | ||
| GenBank mRNA | AF332458.1 | - | 2 - 3 | 545 | 52 | |
| AF422178.1 | - | 2 - 3 | 579 | 52 | ||
| AY034935.1 | - | 3 - 4 | 695 | 52 | ||
| AY113925.1 | - | 2 - 3 | 545 | 52 | ||
More Information
Set Membership: |
|

POM1
Genomic Map