Assay Details
| Gene Symbol: SKIP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331613.1 | NP_001320593.1 | 2 - 3 | 1050 | 79 | |
| NM_100492.4 | NP_563759.1 | 2 - 3 | 1347 | 79 | ||
| GenBank mRNA | AF419599.1 | - | 2 - 3 | 973 | 79 | |
| AY099571.1 | - | 2 - 3 | 991 | 79 | ||
| BT002142.1 | - | 2 - 3 | 907 | 79 | ||
More Information
Set Membership: |
|

SKIP16
Genomic Map