Assay Details
| Gene Symbol: SKIP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331614.1 | NP_001320594.1 | 3 - 4 | 1263 | 96 | |
| NM_100492.4 | NP_563759.1 | 3 - 4 | 1568 | 96 | ||
| GenBank mRNA | AF419599.1 | - | 3 - 4 | 1194 | 96 | |
| AY099571.1 | - | 3 - 4 | 1212 | 96 | ||
| BT002142.1 | - | 3 - 4 | 1128 | 96 | ||
More Information
Set Membership: |
|

SKIP16
Genomic Map