Assay Details
| Gene Symbol: EMB1444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197992.2 | NP_001184921.1 | 1 - 2 | 633 | 78 | |
| NM_001331616.1 | NP_001318931.1 | 1 - 2 | 633 | 78 | ||
More Information
Set Membership: |
|

EMB1444
Genomic Map