Assay Details
| Gene Symbol: ORA59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100497.3 | NP_172106.1 | 1 | 785 | 160 | |
| GenBank mRNA | AK228085.1 | - | 1 | 688 | 160 | |
| BT011001.1 | - | 1 | 687 | 160 | ||
| BT014958.1 | - | 1 | 646 | 160 | ||
More Information
Set Membership: |
|

ORA59
Genomic Map