Assay Details
| Gene Symbol: ACD32.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100526.5 | NP_172134.1 | 2 - 3 | 467 | 98 | |
| GenBank mRNA | BT025652.1 | - | 2 - 3 | 225 | 98 | |
| DQ403189.1 | - | 2 - 3 | 225 | 98 | ||
More Information
Set Membership: |
|

ACD32.1
Genomic Map