Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 10 - 11 | 1668 | 92 | |
| GenBank mRNA | AK227718.1 | - | 10 - 11 | 1653 | 92 | |
| BT006450.1 | - | 10 - 11 | 1623 | 92 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map