Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 11 - 12 | 1731 | 72 | |
| GenBank mRNA | AK227718.1 | - | 11 - 12 | 1716 | 72 | |
| BT006450.1 | - | 11 - 12 | 1686 | 72 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map