Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 1 - 2 | 285 | 68 | |
| GenBank mRNA | AK227718.1 | - | 1 - 2 | 270 | 68 | |
| BT006450.1 | - | 1 - 2 | 240 | 68 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map