Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 2 - 3 | 572 | 65 | |
| GenBank mRNA | AK227718.1 | - | 2 - 3 | 557 | 65 | |
| BT006450.1 | - | 2 - 3 | 527 | 65 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map