Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 5 - 6 | 921 | 155 | |
| GenBank mRNA | AK227718.1 | - | 5 - 6 | 906 | 155 | |
| BT006450.1 | - | 5 - 6 | 876 | 155 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map