Assay Details
| Gene Symbol: AT1G06560 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100535.4 | NP_172143.3 | 9 - 10 | 1590 | 69 | |
| GenBank mRNA | AK227718.1 | - | 9 - 10 | 1575 | 69 | |
| BT006450.1 | - | 9 - 10 | 1545 | 69 | ||
More Information
Set Membership: |
|

AT1G06560
Genomic Map