Assay Details
| Gene Symbol: F2KP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100584.5 | NP_172191.1 | 12 - 13 | 1664 | 122 | |
| GenBank mRNA | AB035288.1 | - | 12 - 13 | 1604 | 122 | |
More Information
Set Membership: |
|

F2KP
Genomic Map