Assay Details
| Gene Symbol: SIRANBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_100588.4 | NP_172194.1 | 1 - 2 | 302 | 76 | |
| GenBank mRNA | AF370491.1 | - | 1 - 2 | 244 | 76 | |
| AF410294.1 | - | 1 - 2 | 246 | 76 | ||
| AY097364.1 | - | 1 - 2 | 154 | 76 | ||
| BT002193.1 | - | 1 - 2 | 154 | 76 | ||
More Information
Set Membership: |
|

SIRANBP
Genomic Map