Assay Details
| Gene Symbol: TRFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331707.1 | NP_001318943.1 | 8 - 9 | 1864 | 116 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRFL2| Gene Symbol: TRFL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001331707.1 | NP_001318943.1 | 8 - 9 | 1864 | 116 | |
Set Membership: |
|