Assay Details
| Gene Symbol: NDR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112952.4 | NP_188696.1 | 1 | 213 | 112 | |
| GenBank mRNA | BT002004.1 | - | 1 | 135 | 112 | |
| BT006267.1 | - | 1 | 107 | 112 | ||
More Information
Set Membership: |
|

NDR1
Genomic Map