Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 15 - 16 | 1924 | 73 | |
| GenBank mRNA | AK226894.1 | - | 15 - 16 | 1774 | 73 | |
| AX765820.1 | - | 15 - 16 | 1717 | 73 | ||
| AY140096.1 | - | 15 - 16 | 1779 | 73 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map