Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 4 - 5 | 928 | 71 | |
| GenBank mRNA | AK226894.1 | - | 4 - 5 | 778 | 71 | |
| AX765820.1 | - | 4 - 5 | 721 | 71 | ||
| AY140096.1 | - | 4 - 5 | 783 | 71 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map