Assay Details
| Gene Symbol: UBP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112954.4 | NP_566666.2 | 6 - 7 | 1154 | 75 | |
| GenBank mRNA | AK226894.1 | - | 6 - 7 | 1004 | 75 | |
| AX765820.1 | - | 6 - 7 | 947 | 75 | ||
| AY140096.1 | - | 6 - 7 | 1009 | 75 | ||
More Information
Set Membership: |
|

UBP14
Genomic Map