Assay Details
| Gene Symbol: AT3G20800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112971.4 | NP_188716.1 | 1 - 2 | 292 | 72 | |
| GenBank mRNA | AY091082.1 | - | 1 - 2 | 223 | 72 | |
| AY122953.1 | - | 1 - 2 | 176 | 72 | ||
More Information
Set Membership: |
|

AT3G20800
Genomic Map