Assay Details
| Gene Symbol: NF-YA9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_112983.5 | NP_566670.1 | 2 - 3 | 408 | 83 | |
| GenBank mRNA | AF412056.1 | - | 2 - 3 | 410 | 83 | |
| AY093971.1 | - | 1 - 2 | 268 | 83 | ||
More Information
Set Membership: |
|

NF-YA9
Genomic Map