Assay Details
| Gene Symbol: BGLU15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337086.1 | NP_001325181.1 | 5 - 6 | 860 | 118 | |
| NM_130008.3 | NP_181973.1 | 10 - 11 | 1609 | 118 | ||
More Information
Set Membership: |
|

BGLU15
Genomic Map