Assay Details
| Gene Symbol: BGLU15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337086.1 | NP_001325181.1 | 3 - 4 | 723 | 88 | |
| NM_130008.3 | NP_181973.1 | 8 - 9 | 1472 | 88 | ||
More Information
Set Membership: |
|

BGLU15
Genomic Map