Assay Details
| Gene Symbol: DAD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337097.1 | NP_001324613.1 | 1 | 1768 | 76 | |
| NM_130045.4 | NP_182008.1 | 1 | 1089 | 76 | ||
| GenBank mRNA | AY074633.1 | - | 1 | 1089 | 76 | |
More Information
Set Membership: |
|

DAD1
Genomic Map