Assay Details
| Gene Symbol: HUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337108.1 | NP_001323852.1 | 14 - 15 | 2311 | 106 | |
| NM_130060.4 | NP_182022.2 | 14 - 15 | 2317 | 106 | ||
| GenBank mRNA | AY081322.1 | - | 14 - 15 | 2256 | 106 | |
| BT010360.1 | - | 14 - 15 | 1912 | 106 | ||
More Information
Set Membership: |
|

HUB1
Genomic Map