Assay Details
| Gene Symbol: HUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130060.4 | NP_182022.2 | 18 - 19 | 2910 | 69 | |
| GenBank mRNA | AY081322.1 | - | 18 - 19 | 2849 | 69 | |
| BT010360.1 | - | 18 - 19 | 2505 | 69 | ||
More Information
Set Membership: |
|

HUB1
Genomic Map