Assay Details
| Gene Symbol: HUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337108.1 | NP_001323852.1 | 9 - 10 | 1731 | 155 | |
| NM_130060.4 | NP_182022.2 | 9 - 10 | 1737 | 155 | ||
| GenBank mRNA | AY081322.1 | - | 9 - 10 | 1676 | 155 | |
| BT010360.1 | - | 9 - 10 | 1332 | 155 | ||
More Information
Set Membership: |
|

HUB1
Genomic Map