Assay Details
| Gene Symbol: SPX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130076.2 | NP_182038.1 | 1 - 2 | 286 | 111 | |
| GenBank mRNA | BT020235.1 | - | 1 - 2 | 285 | 111 | |
| BT020499.1 | - | 1 - 2 | 207 | 111 | ||
More Information
Set Membership: |
|

SPX3
Genomic Map