Assay Details
| Gene Symbol: AT2G45180 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130081.5 | NP_566036.1 | 1 | 379 | 71 | |
| GenBank mRNA | AF424568.1 | - | 1 | 216 | 71 | |
| AK227248.1 | - | 1 | 215 | 71 | ||
| AK230052.1 | - | 657 | 71 | |||
| AK230334.1 | - | 2553 | 71 | |||
| AY085700.1 | - | 1 | 247 | 71 | ||
| BT020290.1 | - | 1 | 205 | 71 | ||
| BT028989.1 | - | 1 | 190 | 71 | ||
More Information
Set Membership: |
|

AT2G45180
Genomic Map