Assay Details
| Gene Symbol: AT2G45620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130124.1 | NP_566048.1 | 1 - 2 | 1287 | 67 | |
| GenBank mRNA | AF360181.1 | - | 1 - 2 | 1287 | 67 | |
| AY039997.1 | - | 1 - 2 | 1208 | 67 | ||
More Information
Set Membership: |
|

AT2G45620
Genomic Map