Assay Details
| Gene Symbol: AT2G45620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130124.1 | NP_566048.1 | 2 - 3 | 1648 | 96 | |
| GenBank mRNA | AF360181.1 | - | 2 - 3 | 1648 | 96 | |
| AY039997.1 | - | 2 - 3 | 1569 | 96 | ||
More Information
Set Membership: |
|

AT2G45620
Genomic Map