Assay Details
| Gene Symbol: AT2G45620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130124.1 | NP_566048.1 | 3 - 4 | 1879 | 87 | |
| GenBank mRNA | AF360181.1 | - | 3 - 4 | 1879 | 87 | |
| AY039997.1 | - | 3 - 4 | 1800 | 87 | ||
More Information
Set Membership: |
|

AT2G45620
Genomic Map