Assay Details
| Gene Symbol: AGL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130127.2 | NP_182089.1 | 3 - 4 | 586 | 85 | |
| GenBank mRNA | DQ446624.1 | - | 3 - 4 | 323 | 85 | |
| M55554.1 | - | 3 - 4 | 426 | 85 | ||
More Information
Set Membership: |
|

AGL6
Genomic Map