Assay Details
| Gene Symbol: AGL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130127.2 | NP_182089.1 | 7 - 8 | 927 | 68 | |
| GenBank mRNA | DQ446624.1 | - | 7 - 8 | 664 | 68 | |
| M55554.1 | - | 7 - 8 | 767 | 68 | ||
More Information
Set Membership: |
|

AGL6
Genomic Map