Assay Details
| Gene Symbol: AGL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337146.1 | NP_001323425.1 | 4 - 5 | 775 | 136 | |
| NM_130128.4 | NP_182090.1 | 4 - 5 | 815 | 136 | ||
| GenBank mRNA | AF385731.1 | - | 4 - 5 | 711 | 136 | |
| AY007726.1 | - | 4 - 5 | 422 | 136 | ||
| AY093967.1 | - | 3 - 4 | 323 | 136 | ||
More Information
Set Membership: |
|

AGL20
Genomic Map