Assay Details
| Gene Symbol: AGL20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337146.1 | NP_001323425.1 | 6 - 7 | 1086 | 111 | |
| NM_130128.4 | NP_182090.1 | 7 - 8 | 1004 | 111 | ||
| GenBank mRNA | AF385731.1 | - | 7 - 8 | 900 | 111 | |
| AY007726.1 | - | 7 - 8 | 611 | 111 | ||
| AY093967.1 | - | 6 - 7 | 512 | 111 | ||
More Information
Set Membership: |
|

AGL20
Genomic Map