Assay Details
| Gene Symbol: CPFTSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130140.3 | NP_566056.1 | 10 - 11 | 1143 | 79 | |
| GenBank mRNA | AF360125.1 | - | 10 - 11 | 1013 | 79 | |
| AJ010820.1 | - | 10 - 11 | 993 | 79 | ||
| AY051026.1 | - | 10 - 11 | 983 | 79 | ||
More Information
Set Membership: |
|

CPFTSY
Genomic Map