Assay Details
| Gene Symbol: CPFTSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130140.3 | NP_566056.1 | 1 - 2 | 469 | 98 | |
| GenBank mRNA | AF360125.1 | - | 1 - 2 | 339 | 98 | |
| AJ010820.1 | - | 1 - 2 | 319 | 98 | ||
| AY051026.1 | - | 1 - 2 | 309 | 98 | ||
More Information
Set Membership: |
|

CPFTSY
Genomic Map