Assay Details
| Gene Symbol: CPFTSY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130140.3 | NP_566056.1 | 9 - 10 | 1078 | 88 | |
| GenBank mRNA | AF360125.1 | - | 9 - 10 | 948 | 88 | |
| AJ010820.1 | - | 9 - 10 | 928 | 88 | ||
| AY051026.1 | - | 9 - 10 | 918 | 88 | ||
More Information
Set Membership: |
|

CPFTSY
Genomic Map