Assay Details
| Gene Symbol: DTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001337151.1 | NP_001324036.1 | 3 - 4 | 1346 | 100 | |
| NM_001337152.1 | NP_001324038.1 | 4 - 5 | 1239 | 100 | ||
| NM_001337153.1 | NP_001324037.1 | 3 - 4 | 1333 | 100 | ||
| NM_001337154.1 | NP_001324039.1 | 3 - 4 | 1046 | 100 | ||
| NM_130146.4 | NP_182107.3 | 3 - 4 | 1328 | 100 | ||
| NM_201965.2 | NP_973694.1 | 2 - 3 | 1431 | 100 | ||
| GenBank mRNA | BT011684.1 | - | 2 - 3 | 581 | 100 | |
| BT012623.1 | - | 2 - 3 | 504 | 100 | ||
| KJ138853.1 | - | 3 - 4 | 927 | 100 | ||
More Information
Set Membership: |
|

DTA2
Genomic Map