Assay Details
| Gene Symbol: AT2G45840 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130147.6 | NP_182108.2 | 5 - 6 | 1335 | 141 | |
| GenBank mRNA | KJ138852.1 | - | 5 - 6 | 1202 | 141 | |
More Information
Set Membership: |
|

AT2G45840
Genomic Map