Assay Details
| Gene Symbol: BAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130151.7 | NP_182112.2 | 5 - 6 | 2011 | 93 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

BAM7| Gene Symbol: BAM7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130151.7 | NP_182112.2 | 5 - 6 | 2011 | 93 | |
Set Membership: |
|