Assay Details
| Gene Symbol: SK20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084596.2 | NP_001078065.1 | 3 - 4 | 822 | 77 | |
| NM_001337163.1 | NP_001325355.1 | 3 - 4 | 1034 | 77 | ||
| NM_001337164.1 | NP_001325353.1 | 2 - 3 | 773 | 77 | ||
| NM_001337165.1 | NP_001325354.1 | 2 - 3 | 773 | 77 | ||
| NM_130158.4 | NP_566058.2 | 3 - 4 | 970 | 77 | ||
More Information
Set Membership: |
|

SK20
Genomic Map