Assay Details
| Gene Symbol: SK20 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084596.2 | NP_001078065.1 | 5 - 6 | 1120 | 106 | |
| NM_001337163.1 | NP_001325355.1 | 5 - 6 | 1332 | 106 | ||
| NM_001337164.1 | NP_001325353.1 | 4 - 5 | 1071 | 106 | ||
| NM_001337165.1 | NP_001325354.1 | 4 - 5 | 1071 | 106 | ||
| NM_130158.4 | NP_566058.2 | 5 - 6 | 1268 | 106 | ||
More Information
Set Membership: |
|

SK20
Genomic Map